Please login to your online account to display your discounted pricing
Fisher Science Education™ Water Quality pH Pens
Highly stable and accurate readings with large LCD display.
Manufacturer: fisher science education  850052F
 View more versions of this product
Catalog No. S96275B
  • / Each
Description & Specifications
Specifications
Accuracy ±0.05
Range 0 to 14
For Use With (Application) Field pH tests
Product Type Water Quality pH Pen
Includes Four button cell batteries and a built in electrode kept moist with a protective cap
  • Automatically calibrates to pH 4, 7 and 10 buffers; can also be adjusted to recognize nonstandard buffers
  • Feature hold function and low battery indicator
  • Automatic three-point calibration and power off