Please login to your online account to display your discounted pricing
Fisher Science Education™ Water Quality pH Pens
Highly stable and accurate readings with large LCD display.
$40.50 - $87.75
Specifications
Product Type Water Quality pH Pen
For Use With (Application) Field pH tests
Includes Four button cell batteries and a built in electrode kept moist with a protective cap
View More Specs
Includes:  Four button-cell batteries and a built-in electrode kept moist with a protective cap
Products
Catalog Number Mfr. No. Accuracy Range Price Quantity    
S96275A
fisher scientific
850051F
±0.2 0 to 14 Each for $48.60
S96275B
fisher science education
850052F
±0.05 0 to 14 Each for $87.75
S96275C
fisher science education
850050F
±0.3 2 to 12 Each for $40.50
Description & Specifications
Specifications
Product Type Water Quality pH Pen
For Use With (Application) Field pH tests
Includes Four button cell batteries and a built in electrode kept moist with a protective cap
  • Automatically calibrates to pH 4, 7 and 10 buffers; can also be adjusted to recognize nonstandard buffers
  • Feature hold function and low battery indicator
  • Automatic three-point calibration and power off
Dimensions: 6.5 × 1.5 × 1.5 in. (165 × 38 × 38mm). Weight: 2.5 oz. (68g).
CE marked. IP65 waterproof.
One year