missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Accumax Lab Technology Variable Micropipets
Automatic tip ejector makes operation fast and convenient.
Manufacturer: Accumax Lab Technology VA800
Includes: Service wrench and instructions
Description
- Ergonomically designed
- Digital display indicates the chosen volume
- Deliver precise and reproducible volumes with ease
- Utilize standard disposable micropipet tips (Except the 1000-5000μL model, S902432K)
- No calibration servicing needed
Warranty
30 days
Specifications
Inaccuracy of +/- 2% for 20uL, or 0.6% for 200uL | |
Ergonomic | |
1μL |
20 to 200 μL | |
Service wrench, instructions | |
30 Days |