missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Accumax Lab Technology Variable Micropipets

Automatic tip ejector makes operation fast and convenient.

Manufacturer:  Accumax Lab Technology VA500

 View more versions of this product

Catalog No. S902432E



Includes: Service wrench and instructions

Description

Description

 

  • Ergonomically designed
  • Digital display indicates the chosen volume
  • Deliver precise and reproducible volumes with ease
  • Utilize standard disposable micropipet tips (Except the 1000-5000μL model, S902432K)
  • No calibration servicing needed

 

Warranty

30 days

Specifications

Specifications

Inaccuracy of +/- 3% for 10uL, or 0.8% for 100uL
Ergonomic
0.5μL
10 to 100 μL
Service wrench, instructions
30 Days
SDS
Documents
Product Certifications
Promotions

Promotions