missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Accumax Lab Technology Variable Micropipets

Automatic tip ejector makes operation fast and convenient.

Manufacturer:  Accumax Lab Technology VA125

 View more versions of this product

Catalog No. S902432A



Includes: Service wrench and instructions

Description

Description

 

  • Ergonomically designed
  • Digital display indicates the chosen volume
  • Deliver precise and reproducible volumes with ease
  • Utilize standard disposable micropipet tips (Except the 1000-5000μL model, S902432K)
  • No calibration servicing needed

 

Warranty

30 days

Specifications

Specifications

Inaccuracy of +/- 12% for 0.25uL, or 2.5% for 2.5uL
Service wrench, instructions
30 Days
0.1 to 2.5 μL
Ergonomic
0.01μL
Education
Documents