Please login to your online account to display your discounted pricing
Fisher Science Education™ University/Laboratory Compound Microscopes
Microscopes incorporate a wide range of features designed for university and laboratory use.
Manufacturer: fisher scientific  160EMD
 View more versions of this product
Catalog No. S71003
  • / Each
Description & Specifications
Specifications
Magnification Power 40X, 100X, 400X, 1000X
Objective Types DIN 4X, 10X, 40X, 100X Achromatic
Nosepiece 4-hole, click stop, reverse angle, ball-bearing mounted
Head Style Monocular
Condenser Type Abbe 1.25 N.A.
Eyepiece 10x, 18mm, with pointer