Please login to your online account to display your discounted pricing
Fisher Science Education™ University/Laboratory Compound Microscopes
Microscopes incorporate a wide range of features designed for university and laboratory use.
$855.10 - $1,672.65
Specifications
Condenser Type Abbe 1.25 N.A.
Magnification Power 40X, 100X, 400X, 1000X
View More Specs
Includes:  Vinyl dust cover
Products
Catalog Number Mfr. No. Head Style Objective Types Price Quantity    
S71003
fisher scientific
160EMD
Monocular DIN 4X, 10X, 40X, 100X Achromatic Each for $855.10
S71003B
fisher scientific
161EMD
Dual-View DIN 4X, 10X, 40X, 100X Achromatic Each for $942.85
S71003A
fisher scientific
160ASCEMD
Monocular DIN 4X, 10X, 40X, 100X ASC Each for $926.65
S71003D
fisher scientific
162EMD
Binocular DIN 4X, 10X, 40X, 100X Achromatic Each for $1,100.25
S71003C
fisher scientific
161ASCEMD
Dual-View DIN 4X, 10X, 40X, 100X ASC Each for $1,014.40
S71003E
fisher scientific
162ASCEMD
Binocular DIN 4X, 10X, 40X, 100X ASC Each for $1,171.80
S71003F
fisher scientific
162PEMD
Binocular DIN 4X, 10X, 40X, 100X Plan Achromatic Each for $1,348.65
S71003G
fisher scientific
163EMD
Trinocular DIN 4X, 10X, 40X, 100X Achromatic Each for $1,434.25
S71003H
fisher scientific
163ASCEMD
Trinocular DIN 4X, 10X, 40X, 100X ASC Each for $1,505.80
S71003J
fisher scientific
163PEMD
Trinocular DIN 4X, 10X, 40X, 100X Plan Achromatic Each for $1,672.65
Description & Specifications
Specifications
Condenser Type Abbe 1.25 N.A.
Magnification Power 40X, 100X, 400X, 1000X
Superb optical clarity and reliable mechanical engineering assure years of top performance.
Optical System
  • Widefield 10X /18mm eyepiece, pointer standard in one eyepiece; accepts optional eyepiece reticle
  • DIN standard 4X (0.10 N.A.), 10X (0.25 N.A.), 40XR (0.65 N.A.) and 100XR oil immersion (1.25 N.A.) objectives
  • All objectives are parfocalled, parcentered and color-coded
  • Quad objective turret
  • Mounted in reverse position to facilitate ease in changing slides
  • Rotates 360° for easy sharing
Illumination
  • 12V 20W halogen with variable rheostat control
  • Illuminated on/off switch, 3–wire grounded cord and plug
  • 0.5A time delay fuse
Focusing
  • Coaxial coarse and fine focusing controls
  • Stage
    • Large 140mm x 135mm (5.5 in. x 5.25 in.) stage
    • Built–in, low profile, ball-bearing mounted mechanical specimen holder with low-position coaxial controls, 70mm by 50mm (x by y) movement with graduations of 0.1mm
    • 1.25 N.A. Abbe condenser has rack & pinion focusing, iris diaphragm, swing out filter holder with clear blue, green and yellow filters supplied
    Frame
    • Rugged cast aluminum, gray enamel finish
    Packaging: Head and frame in molded styrofoam container. Replacement bulb for all models: S74020.
    Lifetime limited warranty, excluding bulb, cord, and fuse