Please login to your online account to display your discounted pricing
Fisher Science Education™ University/Laboratory Compound Microscopes
Microscopes incorporate a wide range of features designed for university and laboratory use.
Manufacturer: fisher scientific  160ASCEMD
 View more versions of this product
Catalog No. S71003A
  • / Each
Description & Specifications
Specifications
Condenser Type Abbe 1.25 N.A.
Objective Types DIN 4X, 10X, 40X, 100X ASC
Magnification Power 40X, 100X, 400X, 1000X
Head Style Monocular