missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Science Education™ Traceable™ Extreme-Accuracy Standards Thermometer

Provides the exact readings required for critical requirements, quality control checks and routine measurements. Fisherbrand Traceable Extreme-Accuracy Standards Thermometers are ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater.

Manufacturer:  Fisher Science Education™ 4242FE

 View more versions of this product

Catalog No. S98177



Includes: Traceable™ Certificate, stainless steel probe, ABS Plastic case, two AA batteries for continuous 1-year always-on monitoring, stand, Velcro™, magnetic strips and wall mount

Description

Description

  • Four digital thermometers
  • Units read 0.00, 25.00, 37.00 or combination of all three
  • Resolution: 0.01°; Accuracy: ±0.05°C within ±2°C of standard
  • Units read in °C and °F
  • Display shows minimum, maximum and current temperatures
  • Stainless steel probe for liquid, air/gas and semisolids
  • Alarm: Two visual (LEDs), two audio alerts
  • Probe tip penetration: 0.03 in.; Diameter: 0.125 in.
  • Stem Length: 6.25 in.; Overall length: 9 in.;
  • Cable length: 5 ft.
  • Display shows minimum/maximum and current temperatures
  • Alarms are programmable in 0.01° increments
  • High-impact; Chemical-resistant ABS plastic case
  • Dimensions : 2.75 x 4.25 x 0.75 in.
  • Weight: 5 oz.

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Specifications

73.4° to 80.6°F
80.6°F
73.4°F
Education
0.12 in.
Measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater
1.9cm
60 in.
107mm
15.9cm
0.01°C
140g
6.9cm
LED
23.0° to 27.0°C
27°C
23°C
DNV, ISO 17025, A2LA
4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9cm)
0.75 in.
Includes: Traceable™ Certificate, stainless steel probe, ABS Plastic case, two AA batteries for continuous 1-year always-on monitoring, stand, Velcro™, magnetic strips and wall mount
6.25 in.
152.4cm
ABS Plastic
5 oz.
2.75 in.
±0.05°C
4.25 in.
Documents