Please login to your online account to display your discounted pricing

Fisher Science Education™ Traceable™ Extreme-Accuracy Standards Thermometer


Provides the exact readings required for critical requirements, quality control checks and routine measurements. Fisher Science Education Traceable Extreme-Accuracy Standards Thermometers are ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater.

Manufacturer: fisher science education™  4241FE

 View more versions of this product

Traceable™ Certificate, stainless steel probe, ABS Plastic case, two AA batteries for continuous 1-year always-on monitoring, stand, Velcro™, magnetic strips and wall mount

Catalog No. S98176

  • / Each
 

Description & Specifications

Specifications

Temperature Range (Metric) -2.0° to +2.0°C
Temperature Range (English) 28.4° to 35.6°F
Length (Metric) Probe 15.9cm
Resolution 0.01°C
Width (Metric) 6.9cm
Certifications/Compliance DNV, ISO 17025, A2LA
Height (English) 0.75 in.
Length (Metric) 10.7cm
Length (Metric) Cable 152.4cm
Weight (Metric) 140g
Display LED
Includes Includes: Traceable™ Certificate, stainless steel probe, ABS Plastic case, two AA batteries for continuous 1-year always-on monitoring, stand, Velcro™, magnetic strips and wall mount
Length (English) 4.25 in.
Length (English) Cable 60 in.
Diameter (English) Probe 0.12 in.
Width (English) 2.75 in.
Length (English) Probe 6.25 in.
For Use With (Application) Measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater
Material ABS Plastic
Weight (English) 5 oz.
Accuracy ±0.05°C
Dimensions (L x W x H) 4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9cm)
Height (Metric) 1.9cm

  • Four digital thermometers
  • Units read 0.00, 25.00, 37.00 or combination of all three
  • Resolution: 0.01°; Accuracy: ±0.05°C within ±2°C of standard
  • Units read in °C and °F
  • Display shows minimum, maximum and current temperatures
  • Stainless steel probe for liquid, air/gas and semisolids
  • Alarm: Two visual (LEDs), two audio alerts
  • Probe tip penetration: 0.03 in.; Diameter: 0.125 in.
  • Stem Length: 6.25 in.; Overall length: 9 in.;
  • Cable length: 5 ft.
  • Display shows minimum/maximum and current temperatures
  • Alarms are programmable in 0.01° increments
  • High-impact; Chemical-resistant ABS plastic case
  • Dimensions : 2.75 x 4.25 x 0.75 in.
  • Weight: 5 oz.

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.