Please login to your online account to display your discounted pricing

Fisher Science Education™ Traceable™ Extreme-Accuracy Standards Thermometer

Provides the exact readings required for critical requirements, quality control checks and routine measurements. Fisher Science Education Traceable Extreme-Accuracy Standards Thermometers are ideal for measuring temperatures in water baths, cuvets, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater.

$145.80 - $157.95

Specifications

Certifications/Compliance DNV, ISO 17025, A2LA
Diameter (English) Probe 0.12 in.
Accuracy ±0.05°C
Dimensions (L x W x H) 4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9cm)
Includes Includes: Traceable™ Certificate, stainless steel probe, ABS Plastic case, two AA batteries for continuous 1-year always-on monitoring, stand, Velcro™, magnetic strips and wall mount
View More Specs
Includes:  Traceable™ Certificate, stainless steel probe, ABS Plastic case, two AA batteries for continuous 1-year always-on monitoring, stand, Velcro™, magnetic strips and wall mount
Products
Catalog Number Mfr. No. Temperature Range (English) Temperature Range (Metric) Price Quantity    

S98176

 
fisher science education™
4241FE
28.4° to 35.6°F -2.0° to +2.0°C Each for $145.80

S98177

 
fisher science education™
4242FE
73.4° to 80.6°F 23.0° to 27.0°C Each for $145.80

S98178

 
fisher science education™
4243FE
95° to 102.2°F 35° to 39°C Each for $145.80

S98179

 
fisher science education™
4244FE
28.4° to 35.6°F, 73.4° to 80.6°F, 95° to 102.2°F -2° to +2°C, 23° to 27°C, 35° to 39°C Each for $157.95
Description & Specifications

Specifications

Certifications/Compliance DNV, ISO 17025, A2LA
Diameter (English) Probe 0.12 in.
Accuracy ±0.05°C
Dimensions (L x W x H) 4.25 x 2.75 x 0.75 in. (10.7 x 6.9 x 1.9cm)
Includes Includes: Traceable™ Certificate, stainless steel probe, ABS Plastic case, two AA batteries for continuous 1-year always-on monitoring, stand, Velcro™, magnetic strips and wall mount
Length (English) Cable 60 in.
Length (English) Probe 6.25 in.
Length (Metric) Cable 152.4cm
Length (Metric) Probe 15.9cm
Resolution 0.01°C
Weight (English) 5 oz.
Weight (Metric) 140g

  • Four digital thermometers
  • Units read 0.00, 25.00, 37.00 or combination of all three
  • Resolution: 0.01°; Accuracy: ±0.05°C within ±2°C of standard
  • Units read in °C and °F
  • Display shows minimum, maximum and current temperatures
  • Stainless steel probe for liquid, air/gas and semisolids
  • Alarm: Two visual (LEDs), two audio alerts
  • Probe tip penetration: 0.03 in.; Diameter: 0.125 in.
  • Stem Length: 6.25 in.; Overall length: 9 in.;
  • Cable length: 5 ft.
  • Display shows minimum/maximum and current temperatures
  • Alarms are programmable in 0.01° increments
  • High-impact; Chemical-resistant ABS plastic case
  • Dimensions : 2.75 x 4.25 x 0.75 in.
  • Weight: 5 oz.

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.