Fisher Science Education™ Student Microscope
Get a closer look at insects, rocks, and other objects
Manufacturer:  Fisher Science Education™ 1851
Catalog No. S190471W
Includes: 10X eyepiece and non-removable 2X objective. Wood case with handle included for safe storage.
Description
Description
Impressive 20X magnification is ideal for viewing three-dimensional objects. No electricity needed; objects can be viewed clearly under normal room light. Designed for the beginner student, images are erect (non-inverted) and operation is simple. Large focusing knobs bring objects into quick focus. Heavy-duty metal body construction holds up to many years of use. Dimensions: 6 1/2 × 3 1/2 × 4 1/2 in..
Specifications
Specifications
Large focusing knobs
Viewing three-dimensional objects
Metal
20X
10X Eyepiece and non removable 2X objective, Wood case with handle included for safe storage