Please login to your online account to display your discounted pricing
Fisher Science Education™ Primary Stereomicroscopes
Popular stereomicroscopes for education and industry.
Manufacturer: fisher scientific  400TBL102EMD
 View more versions of this product
Catalog No. S71004
  • / Each
Description & Specifications
Specifications
Product Type Stereo Microscope
Magnification Power 20X
Objective Types 2X
Eyepiece Paired widefield 10x
Head Style Vertical head
Lighting Type Incidental/Transmitted