Please login to your online account to display your discounted pricing
Fisher Science Education™ Primary+ Compound Microscopes with Coarse Focus Control
Ideal for elementary classroom and field use — compact and versatile.
Manufacturer: fisher scientific  106L
 View more versions of this product
Catalog No. S71000C
  • / Each
Description & Specifications
Specifications
Lighting Type Tungsten
Magnification Power 40X, 100X, 400X
Head Style Monocular
Diaphragm Disc
For Use With (Application) Middle School
Objective Types Achromatic, color-coded 4x, 10x, 40xR objectives
Eyepiece 10x with Pointer