Please login to your online account to display your discounted pricing
Fisher Science Education™ Primary+ Compound Microscopes with Coarse and Fine Focus Control
Ideal for elementary classroom and field use — compact and versatile.
Manufacturer: fisher scientific  109LEMD
 View more versions of this product
Catalog No. S71000E
  • / Each
Description & Specifications
Specifications
Magnification Power 40X, 100X, 400X
Diaphragm Disc
Objective Types Achromatic, Color coded 4x, 10x, 40xR objectives
Eyepiece 10x with Pointer
For Use With (Application) Middle School
Head Style Monocular
Lighting Type Tungsten