Please login to your online account to display your discounted pricing

Fisher Science Education™ Laboratory Benchtop pH Meters


Large, dual LCD shows pH measurement and temperature results simultaneously (with ATC probes).

Manufacturer: fisher science education™  3561710

 View more versions of this product

Catalog No. S90526

  • / Each
 

Description & Specifications

Specifications

Description Economical model. Includes refillable epoxy-body pH electrode.
Model pH Tutor Meter
Includes Meter with Electrode
Range (pH) 0 to 14
Resolution (pH) 0.01
Accuracy (pH) ±0.01
Calibration Points (pH) 3
Resolution (Temperature) 0.1°C
Display Type LCD
Detection Range 0 to 14pH
For Use With (Application) pH, mV, °C
Temperature Range (Metric) 0° to 100°C