missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Science Education™ Laboratory Benchtop pH Meters

Large, dual LCD shows pH measurement and temperature results simultaneously (with ATC probes).

Manufacturer:  Fisher Science Education™ 3561712

 View more versions of this product

Catalog No. S90524



Description

Description

  • pH Mode: 0.00 to 14.00pH with resolution of 0.01 and accuracy of ±0.01pH
  • Temperature Mode (with ATC probe): 0.0 to 100°C with resolution of 0.1°C and accuracy of 0.3°C
  • mV Mode (S90525, S90527 and S90528 only): -1000.0 to +1000.0 with a resolution of 0.1 and an accuracy of 0.1mV within ±199.9mV; 1mV beyond ±199.9mV
  • mV models also feature recorder output
  • Simple push-button standardization with auto-buffer recognition
  • Icons clearly show current meter status
  • Three-point calibration with percent slope calculation
  • Simple user-interface

Warranty

Three years

Specifications

Specifications

LCD
32° to 212°F
±0.01
0.3°C
0 to 14pH
0.01
Meter only
0.1mV
0.1°C
0° to 100°C
0.1mV
3
pH, Temperature
0 to 14
ATC
-10000 to 1000 mV
Documents