missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Science Education™ Laboratory Benchtop pH Meters
Large, dual LCD shows pH measurement and temperature results simultaneously (with ATC probes).
Manufacturer: Fisher Science Education™ 3561730
Description
- pH Mode: 0.00 to 14.00pH with resolution of 0.01 and accuracy of ±0.01pH
- Temperature Mode (with ATC probe): 0.0 to 100°C with resolution of 0.1°C and accuracy of 0.3°C
- mV Mode (S90525, S90527 and S90528 only): -1000.0 to +1000.0 with a resolution of 0.1 and an accuracy of 0.1mV within ±199.9mV; 1mV beyond ±199.9mV
- mV models also feature recorder output
- Simple push-button standardization with auto-buffer recognition
- Icons clearly show current meter status
- Three-point calibration with percent slope calculation
- Simple user-interface
Warranty
Three years
Specifications
LCD | |
0 to 14 pH | |
Meter with Electrode and Swing Arm | |
ATC | |
0 to 14 | |
0.1mV | |
32°F to 212°F |
0°C to 100°C | |
pH, mV, Temperature (°C) | |
pH 510 | |
-10000 to 1000 mV | |
0.01 | |
0.1°C |