Please login to your online account to display your discounted pricing

Fisher Science Education™ pH Meters

Large, dual LCD shows pH measurement and temperature results simultaneously (with ATC probes).

$768.02 - $768.02

Specifications

Accuracy (pH) ±0.01
Detection Range 0 to 14pH
Display Type LCD
Range (pH) 0 to 14
Resolution (pH) 0.01
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Includes Price Quantity    

S90524

 
fisher science education™
WD-35617-12
pH, Temperature Meter only Each for $768.02
Description & Specifications

Specifications

Accuracy (pH) ±0.01
Detection Range 0 to 14pH
Display Type LCD
Range (pH) 0 to 14
Resolution (pH) 0.01
Resolution (Temperature) 0.1°C
Temperature Range (Metric) 0° to 100°C

  • pH Mode: 0.00 to 14.00pH with resolution of 0.01 and accuracy of ±0.01pH
  • Temperature Mode (with ATC probe): 0.0 to 100°C with resolution of 0.1°C and accuracy of 0.3°C
  • mV Mode (S90525, S90527 and S90528 only): -1000.0 to +1000.0 with a resolution of 0.1 and an accuracy of 0.1mV within ±199.9mV; 1mV beyond ±199.9mV
  • mV models also feature recorder output
  • Simple push-button standardization with auto-buffer recognition
  • Icons clearly show current meter status
  • Three-point calibration with percent slope calculation
  • Simple user-interface