Please login to your online account to display your discounted pricing
Fisher Science Education™ Microscope: Reticles
Manufacturer: fisher scientific  96540010EMD
 View more versions of this product
Catalog No. S74037
  • / Each
Description & Specifications
Specifications
Product Type Eyepiece reticle
Size 10mm/100 divisions
For Use With (Equipment) 416 and 417TBL binocular stereoscopic microscope