Please login to your online account to display your discounted pricing

Fisher Science Education™ Mens 4 Gripper Lab Coats


Closure options at right.

Manufacturer: fisher science education  5080LBRGL

 View more versions of this product

Catalog No. S47408A

  • / Each
 

Description & Specifications

Specifications

Material Cotton
Material Polyester
No. of Pockets 3
Closure Type Snap
Item Description Gripper
Length (Metric) 106.6cm
Length (English) 41.5 in.
Color Light Blue
Style Regular
Size Large
Collar Type Notched lapel
Gender Male
Size (English) Chest 44 to 46 in.
Finish Pre-cure durable press