Please login to your online account to display your discounted pricing
Fisher Science Education™ Intermediate WiFi Digital Compound Microscopes
Get high resolution on any budget with these WiFi digital microscopes that transmit live images to iOS™ or Android™ devices.
Manufacturer: fisher scientific  DCX205RLEDEMD
 View more versions of this product
Catalog No. S71014
  • / Each
Description & Specifications
Specifications
Condenser Type Disc 0.65 N.A.
Eyepiece Widefield eyepiece WF10X/18mm
Nosepiece Reversed triple revolving
Magnification Power 4X, 10X, 40X
Objective Types DIN Achromat
Head Style Monocular
Product Type Compound microscope w/floating stage
Includes Triple nosepiece, Corded