Fisher Science Education™ Intermediate+ Stereomicroscopes
Stereomicroscopes offer all the quality, features and versatility of larger stereomicroscopes in a compact size.
$423.75 - $431.85
Specifications
Eyepiece Paired 10X widefield
Head Style Binocular (inclined)
Includes Inclined Head, Corded
Lighting Type Incidental/Transmitted
Product Type Intermediate+ Stereomicroscope
View More Specs
Products 2
Catalog Number Mfr. No. Magnification Power Objective Types Price Quantity  
Catalog Number Mfr. No. Magnification Power Objective Types Price Quantity  
S71006A View Documents Fisher Science Education™
446TBL10EMD
10X, 30X 1X, 3X Each for $423.75
S71006B View Documents Fisher Science Education™
447TBL10EMD
20X, 40X 2X, 4X Each for $431.85
Description
Description
Efficient design lets you select magnification simply by turning the objective turret. Comfortably view your object with 45° eyepieces and 360° rotating head. Positon the microscope in forward or reverse viewing positions.
  • Paired widefield eyepieces with diopter adjustment
  • High-intensity 12V, 10w incidental and transmitted illumination with built-in tension adjustment
  • Rack and pinion focusing
Warranty
Five-year limited
Specifications
Specifications
Paired 10X widefield
Inclined Head, Corded
Intermediate+ Stereomicroscope
Binocular (inclined)
Incidental/Transmitted