Please login to your online account to display your discounted pricing
Fisher Science Education™ Intermediate+ Stereomicroscopes
Stereomicroscopes offer all the quality, features and versatility of larger stereomicroscopes in a compact size.
Manufacturer: fisher scientific  447TBL10EMD
 View more versions of this product
Catalog No. S71006B
  • / Each
Description & Specifications
Specifications
Product Type Intermediate+ Stereomicroscope
Eyepiece Paired 10X widefield
Head Style Binocular (inclined)
Magnification Power 20X, 40X
Lighting Type Incidental/Transmitted
Objective Types 2X, 4X
Includes Inclined Head, Corded