Please login to your online account to display your discounted pricing
Fisher Science Education™ Intermediate+ Stereomicroscopes
Stereomicroscopes offer all the quality, features and versatility of larger stereomicroscopes in a compact size.
Manufacturer: fisher scientific  446TBL10EMD
 View more versions of this product
Catalog No. S71006A
  • / Each
Description & Specifications
Specifications
Product Type Intermediate+ Stereomicroscope
Eyepiece Paired 10X widefield
Objective Types 1X, 3X
Magnification Power 10X, 30X
Head Style Binocular (Inclined)
Lighting Type Incidental/Transmitted
Includes Inclined Head, Corded