Fisher Science Education™ Intermediate Stereomicroscopes
Stereomicroscopes feature rugged design and compact size.
Manufacturer:  Fisher Scientific 453TBL10EMD
 View more versions of this product
Catalog No. S71005B
Specifications Description & Specifications
Specifications
5 years
20X, 40X
Paired widefield 10x
Incidental/Transmitted
Stereo Microscope
Paired, parfocalled, parcentered, achromatic
Binocular (Inclined/Fixed)
Three-Way: Tungsten, Coarded