Please login to your online account to display your discounted pricing
Fisher Science Education™ Intermediate Compound Microscopes: Monocular
Microscopes that set the standard for middle/high school instruction and include all the most commonly specified features.
Manufacturer: fisher scientific  127
 View more versions of this product
Catalog No. S71001
  • / Each
Description & Specifications
Specifications
Eyepiece 10X
For Use With (Application) High School
Head Style Monocular
Magnification Power 40X, 100X, 400X
Lighting Type Tungsten
Diaphragm Disc
Base Style Fixed stage
Objective Types DIN Achromat
Design Type Corded