Please login to your online account to display your discounted pricing
Fisher Science Education™ Intermediate Compound Microscopes: Dual View
Microscopes that set the standard for middle/high school instruction and include all the most commonly specified features.
Manufacturer: fisher scientific  128CLEDMSEMD
 View more versions of this product
Catalog No. S71001F
  • / Each
Description & Specifications
Specifications
Base Style Mechanical stage
Design Type Corded
Diaphragm Disc
Objective Types DIN Achromat
Eyepiece 10X
For Use With (Application) High School
Head Style Dual-View
Lighting Type LED
Magnification Power 40X, 100X, 400X