Please login to your online account to display your discounted pricing

Fisher Science Education™ Foam Cups


Commonly used in the science classroom.

Manufacturer: fisher science education  SC-5001-M

 View more versions of this product

Catalog No. S04197

  • / Pack of 1000 Each
 

Description & Specifications

Specifications

Capacity (English) 12 oz.
Capacity 12 oz., 354.8mL
For Use With (Application) Used in the science classroom
Quantity 1000 Pack
Product Type Cup
Capacity (Metric) 354.8mL
Material Foam