Please login to your online account to display your discounted pricing
Fisher Science Education™ Fixed Minipipets
Easy to use, with precise operation and ergonomic design.
Manufacturer: fisher science education™  AJ-30
 View more versions of this product
Catalog No. S902442D
  • / Each
Description & Specifications
Specifications
Accuracy ±1%
Autoclavable Yes
Compatible Tips Standard disposable micropipet tip
For Use With (Equipment) Standard disposable micropipet tip
Warranty 1 Year
Volume (Metric) 30μL
Compatibility Standard disposable micropipet tip