Please login to your online account to display your discounted pricing
Fisher Science Education™ Fixed Minipipets
Easy to use, with precise operation and ergonomic design.
Manufacturer: fisher science education™  AJ-10
 View more versions of this product
Catalog No. S902442A
  • / Each
Description & Specifications
Specifications
Accuracy ±1%
Autoclavable Yes
Compatible Tips Standard disposable micropipet tip
For Use With (Equipment) Standard disposable micropipet tip
Includes Two step plunger dispenser, smart tip cone design
Warranty 1 Year
Volume (Metric) 10μL
Compatibility Standard disposable micropipet tip