Fisher Science Education™ Fixed Minipipets
Easy to use, with precise operation and ergonomic design.
$61.45 - $61.45
Specifications
Warranty 1 Year
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
S902442D Fisher Science Education™
AJ-30
30μL Each for $61.45
S902442B Fisher Science Education™
AJ-20
20μL Each for $61.45
S902442C Fisher Science Education™
AJ-25
25μL Each for $61.45
S902442E Fisher Science Education™
AJ-35
35μL Each for $61.45
S902442F Fisher Science Education™
AJ-40
40μL Each for $61.45
S902442G Fisher Science Education™
AJ-50
50μL Each for $61.45
S902442H Fisher Science Education™
AJ-75
75μL Each for $61.45
S902442J Fisher Science Education™
AJ-100
100μL Each for $61.45
S902442A Fisher Science Education™
AJ-10
10μL Each for $61.45
S902442K Fisher Science Education™
FA-200
200μL Each for $61.45
Specifications
Description
  • Utilize standard disposable micropipet tips
  • Cost-effective
Warranty
One year
Description & Specifications
Specifications
1 Year