Fisher Science Education™ ChemAlert™ Labeling Kit
Manufacturer: Fisher Science Education™ S47564P
Includes:
- ChemAlert storage plan poster (S47564P)
- List of chemical labeling information for over 500 chemicals (S475648)
- 100 blank NFPA chemical labels sized to fit most chemical containers (S475646)
- Color-coded, pressure-sensitive tape set — one 14 yd. (13m) roll each of blue, red, yellow and white; and two rolls of gray (S68160)
Description
Combines five products for efficient safety labeling of transfer containers, wash bottles and solution bottles.
Specifications
Chemical Storage Plan Poster | |
Chemical hazard | |
1 Ea. |