Fisherbrand™ Anti-Static Static-Away™ Handle Brushes
Designed with the softest hairs available to prevent scratching.
Manufacturer:  Fisherbrand™ 3323FE
 View more versions of this product
Catalog No. S98212
Description
Description
Ideal for dusting balances, cuvets, microscope slides, optics and instrument and computer screens.
  • Removes static charges as it brushes
  • Conductive composite handle
  • Long handle allows access to difficult places
  • Eliminates the possibility of a hand touching
Specifications
Specifications
Education
Ideal for dusting balances, cuvets, microscope slides, optics and instrument/computer screens, designed with the softest hairs available to prevent scratching
6 in.
15.24cm
14.17g
Anti Static Brush
3 in.
7.62cm
0.5 oz.
Documents