Please login to your online account to display your discounted pricing
Fisher Science Education™ Advanced Stereomicroscopes
Suitable for high-school or college classroom and laboratory use.
Manufacturer: fisher science education™  S90014B
 View more versions of this product
Catalog No. S90014B
  • / Each
Description & Specifications
Specifications
Warranty 5 Years
Axis Type Dual coarse and slip clutch mechanism
Design Type Locked in stage clip
Base Style Solid
Magnification Power 10x, 30x
Objective Types 10x, 30x fixed
Quantity 1
Eyepiece 10X
Lighting Type Dual Tungsten
Includes Dust cover, black and white stage plate
Grade High school and college
Head Style Binocular