Please login to your online account to display your discounted pricing
Fisher Science Education™ Advanced+ Stereomicroscopes
Stereomicroscopes that are ideal for college instruction.
Manufacturer: fisher scientific  431430PHF10EMD
 View more versions of this product
Catalog No. S71009A
  • / Each
Description & Specifications
Specifications
Product Type Stereo Microscope
Magnification Power 20X, 40X
Head Style Binocular
Objective Types 2X, 4X
Lighting Type Incidental/Transmitted