Please login to your online account to display your discounted pricing
Fisher Science Education™ Advanced+ Stereomicroscopes
Stereomicroscopes that are ideal for college instruction.
Manufacturer: fisher scientific  420420AHF10EMD
 View more versions of this product
Catalog No. S71008
  • / Each
Description & Specifications
Specifications
Product Type Stereo Zoom Microscope
Includes 80mm frosted glass and black/white contrast plate
Objective Types Zoom
Magnification Power 10X to 40X
Lighting Type Incidental/Transmitted
Head Style Binocular