Fisher Science Education™ Advanced+ Stereomicroscopes
Stereomicroscopes that are ideal for college instruction.
$388.25 - $1,111.85
Specifications
Lighting Type Incidental/Transmitted
View More Specs
Includes: Rubber eyeshields, vinyl dustcover
Products 5
Catalog Number Mfr. No. Magnification Power Objective Types Head Style Product Type Price Quantity  
Catalog Number Mfr. No. Magnification Power Objective Types Head Style Product Type Price Quantity  
S71008 View Documents Fisher Science Education™
420420AHF10EMD
10X to 40X Zoom Binocular Stereo Zoom Microscope Each for $966.05
S71009 View Documents Fisher Science Education™
430430PHF10EMD
10X, 30X 1X, 3X Binocular Stereo Microscope Each for $631.25
S71008A View Documents Fisher Science Education™
420T420AHF10EMD
10X to 40X Zoom Trinocular Stereo Zoom Microscope Each for $1,111.85
S71009A View Documents Fisher Science Education™
431430PHF10EMD
20X, 40X 2X, 4X Binocular Stereo Microscope Each for $388.25
S71008B View Documents Fisher Science Education™
420T430PHF10EMD
10X to 40X Zoom Trinocular Stereo Zoom Microscope Each for $1,111.85
Description
Description
Optical System
  • Paired widefield 10X eyepieces are 45° inclined, with dual diopter adjustments, interpupillary adjustment from 55mm to 75mm
  • Eyepieces equipped with reticle holder/retainer to accept reticle with 23mm O.D.
  • Viewing head rotates 360°
  • Achromatic, color corrected lenses
Illumination
  • Top light (incidental) is halogen, bi-pin with adjustable beam and variable intensity control
  • Bottom light (transmitted) is fluorescent
  • 4-position light control: top only/bottom only/top and bottom/off
Focusing
  • Heavy-duty rack and pinion focusing with slip clutch
  • Tension adjustment eliminates focus 'drift'
Stage
  • Includes two 80mm stage plates: frosted glass and reversible black/white plastic contrast plate
  • Blue filter for transmitted light
  • Locked-on stage clips
Frame
  • Professional, all-metal frame, gray enamel finish
  • Heavy-duty construction
Warranty
Lifetime limited warranty excluding bulb, cord and fuse
Specifications
Specifications
Incidental/Transmitted