Please login to your online account to display your discounted pricing
Fisher Science Education™ Advanced Digital Compound Microscopes
Superior in design and performance with wide range of features designed for university and laboratory use.
Manufacturer: fisher scientific  DC5163EMD
 View more versions of this product
Catalog No. S71012
  • / Each
Description & Specifications
Specifications
For Use With (Application) University and laboratory use
Camera Type Digital
Condenser Type Abbe 1.25 N.A.
Magnification Power 40X, 100X, 400X, 1000X