Please login to your online account to display your discounted pricing
Fisher Science Education™ Advanced Compound Microscopes
Ideal for teaching microscopy to students of varying education levels.
Manufacturer: fisher scientific  212RLEDEMD
 View more versions of this product
Catalog No. S71002F
  • / Each
Description & Specifications
Specifications
Head Style Monocular
Eyepiece 10X
For Use With (Application) High school
Design Type Cordless
Lighting Type LED
Base Style Mechanical stage
Objective Types DIN Achromat
Diaphragm Iris
Magnification Power 40X, 100X, 400X, 1000X
Model 212-RLED
Includes Pointer; 4 Hole Objective Turret