Please login to your online account to display your discounted pricing
Fisher Science Education™ Advanced Compound Microscopes
Ideal for teaching microscopy to students of varying education levels.
Manufacturer: fisher scientific  211RLEDEMD
 View more versions of this product
Catalog No. S71002D
  • / Each
Description & Specifications
Specifications
Base Style Mechanical stage
Diaphragm Iris
Eyepiece 10X
For Use With (Application) High school
Head Style Monocular
Model 211-RLED
Design Type Cordless
Lighting Type LED
Includes Pointer; 4 Hole Objective Turret
Magnification Power 40X, 100X, 400X
Objective Types DIN Achromat