Please login to your online account to display your discounted pricing
Fisher Science Education™ Advanced Compound Microscopes
Ideal for teaching microscopy to students of varying education levels.
Manufacturer: fisher scientific  210RLEDEMD
 View more versions of this product
Catalog No. S71002B
  • / Each
Description & Specifications
Specifications
Model 210-RLED
Base Style Mechanical stage
Design Type Cordless
Magnification Power 40X, 100X, 400X
Diaphragm Iris
Objective Types DIN Achromat
Eyepiece 10X
For Use With (Application) High school
Head Style Monocular
Lighting Type LED
Includes Pointer; 4 Hole Objective Turret