Please login to your online account to display your discounted pricing
Fisher Science Education™ Advanced Compound Microscopes
Ideal for teaching microscopy to students of varying education levels.
Manufacturer: fisher scientific  210EMD
 View more versions of this product
Catalog No. S71002A
  • / Each
Description & Specifications
Specifications
Eyepiece 10X
Diaphragm Disc (0.65 N.A.)
Head Style Monocular
Objective Types DIN Achromat
For Use With (Application) High school
Lighting Type LED
Design Type Corded
Magnification Power 40X, 100X, 400X
Base Style Fixed stage
Includes Pointer; 3 Hole Objective Turret