Please login to your online account to display your discounted pricing
Fisher Science Education™ 160 Series Compound Microscopes
Plan phase lenses provide additional image contrast for live specimen.
Manufacturer: fisher scientific  163PHEMD
 View more versions of this product
Catalog No. S71003L
  • / Each
Description & Specifications
Specifications
Head Style Trinocular
Magnification Power 40X, 100X, 400X, 1000X
Objective Types Plan phase
Condenser Type Abbe 1.25 N.A.