Please login to your online account to display your discounted pricing
Fisher Safety™ Wall-Mounted Eye/Facewashes
Mounted on 0.5 in. (1.27cm) galvanized steel supply arm protected with yellow polyurethane coating
$498.28 - $498.28
Products
Catalog Number Mfr. No. Price Quantity    
19-120-2305
bradley
S19220FW2305
Each for $498.28
Description & Specifications
  • Both models have 10 in. (25.4cm) bowls and exceed minimum water flow of 3gpm (11.35L/min.) at 30psi
  • Activated by highly visible yellow PVC or stainless-steel push handle
  • ABS plastic perforated disc sprayheads gently cleanse eyes and face
  • Sprayhead assemblies include flow control and pop-off protective sprayhead dust covers
  • Stay-open valves are 1/2 in. IPS chrome-plated brass
  • With heavy-gauge aluminum wall bracket and tailpiece, dome-type strainer, and 1.25 in. (3.18cm) IPS female fitting with tailpiece
SEI certified to comply with ANSI™ Z358.1-2004.