Fisherbrand™ Handheld Drench Hoses with Pop-Off Covers
Soft spray for cleansing eyes and face
Manufacturer:  Fisherbrand™ S19430A
 View more versions of this product
Catalog No. 19-120-2313
Description
Description
  • Single vertical perforated ABS plastic sprayhead with protective pop-off cover; choose from deck- or wall-mount options
  • Integral flow control assures safe, balanced flow exceeding 3 gpm at 30psi
  • Chrome-plated valve with extended reinforced handle remains open once handle is squeezed
  • Features 8 ft. long (2.4m) yellow reinforced thermoplastic hose (250psi burst strength) with 0.38 in. (0.95cm) IPS male thread with removable 0.5 in. (1.27cm) IPS reducing coupling; 0.5 in. IPS supply inlet
Specifications
Specifications
250psi
3 gpm at 30psi
Mounting hardware, single ABS head, Thermoplastic hose with IPS male thread and removable IPS reducing coupling
0.939cm
2.4m
Drench Hose
ANSI Z358.1-2004
Soft spray for cleansing eyes and face
0.37 in.
8 ft.
Wall