Fisher Safety™ Handheld Drench Hoses with Pop-Off Covers
Soft spray for cleansing eyes and face
$321.91 - $393.28
Includes: Deck-mount drench hose: mounting hardware and undercounter hose guides. Wall-mount drench hose: mounting hardware.
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
19-120-2312 Bradley
S19460
Each for $393.28
19-120-2313 Bradley
S19430A
Each for $321.91
Specifications
Description
  • Single vertical perforated ABS plastic sprayhead with protective pop-off cover; choose from deck- or wall-mount options
  • Integral flow control assures safe, balanced flow exceeding 3 gpm at 30psi
  • Chrome-plated valve with extended reinforced handle remains open once handle is squeezed
  • Features 8 ft. long (2.4m) yellow reinforced thermoplastic hose (250psi burst strength) with 0.38 in. (0.95cm) IPS male thread with removable 0.5 in. (1.27cm) IPS reducing coupling; 0.5 in. IPS supply inlet
Certifications
SEI certified to comply with ANSI™ Z358.1-2004.