missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Healthcare™ PROTOCOL™ Wright Stain Solution

$144.52 - $308.45

Specifications

Boiling Point 65°C
Format Liquid
Color Dark Purple
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
23-264980
missing translation for 'viewDocuments' Promotion Details
Fisher Healthcare™
264980
16 oz.
Each for $144.52
Case of 12 Each for $1,734.24
 
23-200733
missing translation for 'viewDocuments' Promotion Details
Fisher Healthcare™
200733
1L
Each for $153.55
Case of 12 Each for $1,842.60
 
23-264981
missing translation for 'viewDocuments' Promotion Details
Fisher Healthcare™
264981
1 gal.
Each for $308.45
Case of 4 Each for $1,233.80
 
Description

Description

Specifications

Specifications

65°C
Liquid
Dark Purple
SDS
Documents

Documents