Fisher HealthCare™ PROTOCOL™ SecureMount™
Compatible with both xylene and xylene substitutes and does not crack or discolor with age
$70.20 - $82.45
Specifications
Applications Compatible with both Xylene and Xylene substitutes and does not crack or discolor with age
Volume (English) 16 oz.
Volume (Metric) 475mL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
23-022208 Fisher HealthCare™
022208
PROTOCOL™ SecureMount™, Low Viscosity Each for $70.20
Case of 2 Each for $140.40
23-022210 Fisher HealthCare™
022210
PROTOCOL™ SecureMount™, High Viscosity Each for $82.45
Case of 2 Each for $164.90
Specifications
Description
  • Clear, toluene-based mounting media formulated for superior tissue specimen adherence to slides
  • Dries quickly, allowing slide examination soon after application
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Compatible with both Xylene and Xylene substitutes and does not crack or discolor with age
16 oz.
475mL