missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Healthcare™ PROTOCOL™ Color-Coded Prefilled Formalin Containers
Prefilled with 10% neutral buffered formalin
Supplier: Fisher Healthcare™ 316154
Description
- Color-coded screw cap with leakproof O-ring
- Autoclavable polypropylene
- Paper labels to indicate date, doctor, patient name or ID, and specimen type
- Graduated
Order Info
The 20/10, 40/20 and 60/30mL containers are packaged in shrink wrapped trays to allow for ease of use, handling and storage.
Specifications
Yes | |
Autoclavable | |
Red | |
Paper |
Polypropylene | |
40 mL, 20 mL Formalin fill | |
Color-coded Cap with Leakproof O-Ring |